Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPF8 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB290347 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290347 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290347 Supplier Novus Biologicals Supplier No. NBP222274PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRPF8 Polyclonal antibody specifically detects PRPF8 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen PRPF8
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias apoptosis-regulated protein 1, apoptosis-regulated protein 2, p220, precursor mRNA processing protein, pre-mRNA-processing-splicing factor 8, Prp8, PRP8 homolog, PRP8 pre-mRNA processing factor 8 homolog (S. cerevisiae), PRP8 pre-mRNA processing factor 8 homolog (yeast), PRP8,220 kDa U5 snRNP-specific protein, PRPC8HPRP8, RP13, Splicing factor Prp8, U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 2285 and 2335 of human Pre-mRNA-Processing Factor 8 using the numbering given in entry NP_006436.3 (GeneID 10594).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 10594
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.