Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPF8 Rabbit anti-Human, Mouse, Rat, Clone: 4I1Q8, Novus Biologicals™
SDP

Catalog No. NB167971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167971 100 μg
NB167972 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167971 Supplier Novus Biologicals Supplier No. NBP316440100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PRPF8 Monoclonal antibody specifically detects PRPF8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PRPF8
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4I1Q8
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias apoptosis-regulated protein 1, apoptosis-regulated protein 2, p220, precursor mRNA processing protein, pre-mRNA-processing-splicing factor 8, Prp8, PRP8 homolog, PRP8 pre-mRNA processing factor 8 homolog (S. cerevisiae), PRP8 pre-mRNA processing factor 8 homolog (yeast), PRP8,220 kDa U5 snRNP-specific protein, PRPC8HPRP8, RP13, Splicing factor Prp8, U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 2200-2335 of human PRPF8 (Q6P2Q9).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 10594
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.