Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRR11 Antibody (OTI2A10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP273677A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273677A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273677A30 Supplier Novus Biologicals Supplier No. NBP273677AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRR11 Monoclonal specifically detects PRR11 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PRR11
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2A10
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ11029, proline rich 11, proline-rich protein 11, transcription repressor of MHCII
Gene Symbols PRR11
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 125-360 of human PRR11 (NP_060774) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55771.0
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.