Learn More
Description
Specifications
Specifications
| Antigen | PRR7 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | TRAP3/10 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
| Gene Alias | MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein |
| Gene Symbols | PRR7 |
| Host Species | Mouse |
| Immunogen | Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
