Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRR7 Antibody (TRAP3/10), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB358920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358920 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358920 Supplier Novus Biologicals Supplier No. NBP130424MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRR7 Monoclonal antibody specifically detects PRR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen PRR7
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone TRAP3/10
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein
Host Species Mouse
Immunogen Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80758
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.