Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRRC2B Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB051968 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB051968 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB051968 Supplier Novus Biologicals Supplier No. NBP298065H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRRC2B Polyclonal specifically detects PRRC2B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PRRC2B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias BAT2L, DKFZp781F05101, HLA-B associated transcript 2-like 1, HLA-B-associated transcript 2-like 1, LQFBS-1, MGC10526, proline-rich coiled-coil 2B, proline-rich coiled-coil protein 2B, protein BAT2-like 1, protein PRRC2B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PRRC2B fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84726
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.