Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRRG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159906
Description
PRRG1 Polyclonal specifically detects PRRG1 in Human samples. It is validated for Western Blot.Specifications
PRRG1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
PRGP1TMG1, proline rich Gla (G-carboxyglutamic acid) 1, Proline-rich gamma-carboxyglutamic acid protein 1, proline-rich Gla (G-carboxyglutamic acid) polypeptide 1, Proline-rich Gla protein 1, transmembrane gamma-carboxyglutamic acid protein 1 | |
Rabbit | |
Affinity purified | |
RUO | |
5638 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
O14668 | |
PRRG1 | |
Synthetic peptides corresponding to PRRG1(proline rich Gla (G-carboxyglutamic acid) 1) The peptide sequence was selected from the N terminal of PRRG1. Peptide sequence MGRVFLTGEKANSILKRYPRANGFFEEIRQGNIERECKEEFCTFEEAREA. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Canine: 92%; Mouse: 92%; Rabbit: 92%; Rat: 92%; Xenopus: 91%; Guinea pig: 85%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction