Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRRG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PRRG1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PRRG1 Polyclonal specifically detects PRRG1 in Human samples. It is validated for Western Blot.Specifications
PRRG1 | |
Polyclonal | |
Rabbit | |
O14668 | |
5638 | |
Synthetic peptides corresponding to PRRG1(proline rich Gla (G-carboxyglutamic acid) 1) The peptide sequence was selected from the N terminal of PRRG1. Peptide sequence MGRVFLTGEKANSILKRYPRANGFFEEIRQGNIERECKEEFCTFEEAREA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
PRGP1TMG1, proline rich Gla (G-carboxyglutamic acid) 1, Proline-rich gamma-carboxyglutamic acid protein 1, proline-rich Gla (G-carboxyglutamic acid) polypeptide 1, Proline-rich Gla protein 1, transmembrane gamma-carboxyglutamic acid protein 1 | |
PRRG1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title