Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRSS27 Monoclonal Antibody (04), Invitrogen™
Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | PRSS27 |
---|---|
Clone | 04 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543536
|
Invitrogen™
MA543536 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543032
|
Invitrogen™
MA543032 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human Pancreasin/Marapsin/PRSS27
Pancreasin is a pancreatic tryptic serine peptidase that cleaves peptides after an arginine residue., Sep 2003].Specifications
PRSS27 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
CAP-2; CAPH2; channel-activating protease 2; channel-activating protease 2-like protein; EC 3.4.21; EC 3.4.21.4; EC 3.4.21.43; marapsin; Mpn; MPNMarapsin; Pancreasin; protease, serine 27; PRSS27; Serine protease 27; UNQ1884/PRO4327 | |
PRSS27 | |
IgG1 | |
Protein A | |
Antibody |
04 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9BQR3 | |
83886 | |
Recombinant Human Pancreasin/Marapsin/PRSS27 Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
PRSS27 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title