Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PRT 4165

Click to view available options
Quantity:
10 mg
50 mg
Description
PRT 4165 is an inhibitor of Bmi1/Ring1A, subunits of the polycomb repressive complex 1 (PRC1); inhibits self-ubiquitination (IC50 = 3.9 μM) but does not increase cellular levels of either subunit. Prevents Bmi1/Ring1A-mediated ubiquitination and drug-induced degradation of topoisomerase 2α (Top2α). Also shown to inhibit the in vitro E3 ubiquitin ligase activity of RNF2 and a Bmi1/RNF2 complex, inhibiting H2A/H2AX ubiquitination. Blocks polycomb repressor complex (PRC) 1-mediated histone H2A ubiquitination in vitro.
Specifications
Specifications
Quantity | 10 mg |
Product Type | PRT 4165 |
Purity | >99% |
Molecular Weight (g/mol) | 235.24 |
Inhibitors | PRT 4165 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction