Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUJIFILM Wako Limulus PS Single Test

Catalog No. 12333052 Shop All FujiFilm Products
Click to view available options
Product Type:
Single Test

Contains endotoxin-specific single test vials and an affinity resin suspension designed to overcome any product interference by adsorbing potential endotoxin in samples while washing away inhibitory components.

  • Endotoxin-specific reagent avoids false positive results from glucans
  • Kit comes in single-test configuration to avoid contamination
  • KTA quantitation performed on Toxinometer™
  • Allows endotoxin-specific measurement in samples that typically have inhibitory components:
    • Ethanol
    • Silicone oil
    • Soybean oil
    • Olive oil
    • Stearic acid
    • Egg yolk lecithin
    • Oil adjuvanted vaccine
    • Albumin products
    • Coagulation factor products (factor 8)
    • Interferon formulation
    • Antithrombin III
    • Immunoglobulin product
    • Other ethanol-soluble samples
TRUSTED_SUSTAINABILITY

Specifications

Form Lyophilized, Liquid
Product Type Single Test
Fill Volume 200 μL
For Use With (Equipment) Toxinometer™
Includes 20 single test vials of lysate, PyroSep™ resin suspension, LAL reconstitution solution, Wash solution, Sample diluent, 20 glass capillary columns
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.