Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PSB 069
GREENER_CHOICE

Catalog No. 257310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-731-0 10 mg
25-735-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-731-0 Supplier Tocris Bioscience™ Supplier No. 2573/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-selective NTPDase inhibitor

Non-selective nucleoside triphosphate diphosphohydrolase (NTPDase) inhibitor. Reported to inhibit rat NTPDases 1, 2 and 3 with similar potencies.

Chemical Identifiers

CAS 78510-31-3
Molecular Formula C20H12ClN2NaO5S

Specifications

Quantity 10 mg
Formula Weight 450.83
Purity >98%
Molecular Weight (g/mol) 450.825
Chemical Name or Material PSB 069
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.