Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PSB 10 hydrochloride
GREENER_CHOICE

Catalog No. 201010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-101-0 10 mg
20-105-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-101-0 Supplier Tocris Bioscience™ Supplier No. 2010/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, highly selective hA3 receptor antagonist/inverse agonist

Potent and highly selective antagonist for the human adenosine A3 receptor, with low affinity for the rat A3 receptor (Ki values are 0.44 and > 17000 nM respectively). Displays > 3800-fold selectivity over human A1, A2A and A2B receptors (Ki values are 4.1, 3.3 and 30 μM respectively) and > 1800-fold selectivity over rat A1 and A2A receptors. Acts as an inverse agonist in the [35S]GTPγS binding assay in hA3-CHO cells (IC50 = 4 nM). Produces thermal hyperalgesia in mice in vivo.

Chemical Identifiers

CAS 591771-91-4
Molecular Formula C16H15Cl4N5O

Specifications

Quantity 10 mg
Formula Weight 435.14
Purity >98%
Molecular Weight (g/mol) 435.13
Chemical Name or Material PSB 10 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.