Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PSB 10 hydrochloride
GREENER_CHOICE

Catalog No. p-4784363
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, highly selective hA3 receptor antagonist/inverse agonist

Potent and highly selective antagonist for the human adenosine A3 receptor, with low affinity for the rat A3 receptor (Ki values are 0.44 and > 17000 nM respectively). Displays > 3800-fold selectivity over human A1, A2A and A2B receptors (Ki values are 4.1, 3.3 and 30 μM respectively) and > 1800-fold selectivity over rat A1 and A2A receptors. Acts as an inverse agonist in the [35S]GTPγS binding assay in hA3-CHO cells (IC50 = 4 nM). Produces thermal hyperalgesia in mice in vivo.

Chemical Identifiers

CAS 591771-91-4
Molecular Formula C16H15Cl4N5O

Specifications

Quantity 10 mg
Formula Weight 435.14
Purity >98%
Molecular Weight (g/mol) 435.13
Chemical Name or Material PSB 10 hydrochloride
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.