Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PSB 1115
GREENER_CHOICE

Catalog No. p-4784367
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective human A2B receptor antagonist; water-soluble

Highly selective, water-soluble, human A2B adenosine receptor antagonist. Ki values are 53.4, > 10000 and > 10000 nM at human A2B, A1 and A3 receptors respectively. Also selective versus rat A1 and A2A receptors (Ki values are 2200 and 24000 nM respectively). Produces potent analgesic effects in vivo.

Chemical Identifiers

CAS 152529-79-8
Molecular Formula C14H14N4O5S

Specifications

Quantity 10 mg
Formula Weight 350.35
Purity >98%
Molecular Weight (g/mol) 350.349
Chemical Name or Material PSB 1115
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.