Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PSB 603
GREENER_CHOICE

Catalog No. p-4784371
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective A2B antagonist

Adenosine A2B receptor antagonist that displays > 17000-fold selectivity over other adenosine receptors (Ki values are 0.553, > 10000, > 10000 and > 10000 nM for A2B, A1, A2A and A3 receptors respectively).

Chemical Identifiers

CAS 1092351-10-4
Molecular Formula C24H23ClN6O4S

Specifications

Quantity 10 mg
Formula Weight 529.01
Purity >98%
Molecular Weight (g/mol) 526.996
Chemical Name or Material PSB 603
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.