Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSD-95 Antibody (6G6-1C9), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB300556G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300556G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300556G Supplier Novus Biologicals Supplier No. NB300556G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSD-95 Monoclonal specifically detects PSD-95 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen PSD-95
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 6G6-1C9
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15
Gene Symbols DLG4
Host Species Mouse
Immunogen Purified recombinant rat PSD-95.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1742
Test Specificity Detects Post Synaptic Density 95 kDa (PSD-95) from rat tissues.
Target Species Human, Mouse, Rat, Invertebrate, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.