Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSD-95 Antibody (7E3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP212872B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212872B 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212872B Supplier Novus Biologicals Supplier No. NBP212872B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSD-95 Monoclonal specifically detects PSD-95 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen PSD-95
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 7000
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Alias discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15
Gene Symbols DLG4
Host Species Mouse
Immunogen Recombinant rat PSD-95
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1742
Test Specificity Detects ∼100 kDa. An additional protein of >100 kDa is also detected. Additional cross-reactive bands are detected at ∼75 kDa and 50 kDa in rat and mouse samples.
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.