Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSD3 Antibody, Novus Biologicals™
SDP

Catalog No. NB394879 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB394879 25 μL
NBP255678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB394879 Supplier Novus Biologicals Supplier No. NBP25567825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSD3 Polyclonal specifically detects PSD3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PSD3
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias ADP-ribosylation factor guanine nucleotide factor 6, DKFZp761K1423, EFA6RHepatocellular carcinoma-associated antigen 67, Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6, HCA67Pleckstrin homology and SEC7 domain-containing protein 3, KIAA0942, PH and SEC7 domain-containing protein 3, pleckstrin and Sec7 domain containing 3
Gene Symbols PSD3
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FSAPPFPAAIGSQKKFSRPLLPATTTKLSQEEQLKSHESKLKQITTELAEHRSYPPDKKVKAKDVDEYKLKDHYLEFEKTRYEMYVSILKEGGKELLSNDESEAAGLKKSHSSPSLNPDTSPITAKVKRNVSERKDHRPETPSIKQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23362
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.