Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSD93 Rabbit anti-Human, Mouse, Rat, Clone: 0G4O6, Novus Biologicals™
SDP

Catalog No. NB166793 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166793 100 μg
NB166794 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166793 Supplier Novus Biologicals Supplier No. NBP315849100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PSD93 Monoclonal antibody specifically detects PSD93 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen PSD93
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 0G4O6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Chapsyn-110, discs, large homolog 2 (Drosophila), discs, large homolog 2, chapsyn-110, discs, large homolog 2, chapsyn-110 (Drosophila), disks large homolog 2, DKFZp781D1854, DKFZp781E0954, FLJ37266, MGC131811, Postsynaptic density protein PSD-93, PSD-93, PSD93,110kDa
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 600-700 of human PSD93 (Q15700).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1740
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.