Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSENEN/PEN2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224611SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224611SS 0.025 mg
NBP224611 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224611SS Supplier Novus Biologicals Supplier No. NBP224611SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSENEN/PEN2 Polyclonal specifically detects PSENEN/PEN2 in Human, Mouse, Rat, Bovine, Canine, Equine, Opossum samples. It is validated for Western Blot.

Specifications

Antigen PSENEN/PEN2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-7 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9NZ42
Gene Alias gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2
Gene Symbols PSENEN
Host Species Rabbit
Immunogen A portion of amino acid 1 to 50 of human Presenilin enhancer protein 2 was used as immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Growth and Development, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55851
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse, monkey, mouse, rat and 93% homologous in cow.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Opossum
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.