Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSENEN/PEN2 Antibody, Novus Biologicals™
SDP

Catalog No. NB300227SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300227SS 0.025 mL
NB300227 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300227SS Supplier Novus Biologicals Supplier No. NB300227SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

PSENEN/PEN2 Polyclonal specifically detects PSENEN/PEN2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PSENEN/PEN2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2500, Immunoprecipitation 1:50-1:100
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. Q9NZ42
Gene Alias gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2
Gene Symbols PSENEN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of human PEN2 (within residues 50-101). [UniProt# Q9NZ42]
Molecular Weight of Antigen 12 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55851
Test Specificity Human and mouse neuroblastoma (N2a) cells.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.