Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSENEN/PEN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$181.90 - $487.50
Specifications
Antigen | PSENEN/PEN2 |
---|---|
Dilution | Western Blot 1:1000-1:2500, Immunoprecipitation 1:50-1:100 |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300227SS
![]() |
Novus Biologicals
NB300227SS |
0.025 mL |
Each for $181.90
|
|
|||||
NB300227
![]() |
Novus Biologicals
NB300227 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
PSENEN/PEN2 Polyclonal specifically detects PSENEN/PEN2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
PSENEN/PEN2 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Growth and Development, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
Whole antisera with 0.05% Sodium Azide | |
gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2 | |
PSENEN | |
IgG | |
Unpurified | |
Human and mouse neuroblastoma (N2a) cells. |
Western Blot 1:1000-1:2500, Immunoprecipitation 1:50-1:100 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse | |
Q9NZ42 | |
55851 | |
A synthetic peptide corresponding to the C-terminus of human PEN2 (within residues 50-101). [UniProt# Q9NZ42] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
12 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title