Learn More
Pseudomonas Aeruginosa Antibody (1010/287), DyLight 650, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Pseudomonas Aeruginosa |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1010/287 |
| Conjugate | DyLight 650 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Pseudomonas aeruginosa 9 |
| Host Species | Mouse |
| Immunogen | P. aeruginosa serotype 6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.