Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudomonas Aeruginosa Antibody (1010/287), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP45176646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP45176646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP45176646 Supplier Novus Biologicals Supplier No. NBP145176JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PseudomonasAeruginosa
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1010/287
Conjugate Janelia Fluor 646
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Pseudomonas aeruginosa 9
Host Species Mouse
Immunogen P. aeruginosa serotype 6
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity AERUGINOSA
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.