Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudomonas Aeruginosa Antibody (1200/472), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB380667 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380667 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380667 Supplier Novus Biologicals Supplier No. NBP308420AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pseudomonas Aeruginosa Monoclonal antibody specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA

Specifications

Antigen Pseudomonas Aeruginosa
Applications ELISA
Classification Monoclonal
Clone 1200/472
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Pseudomonas aeruginosa 9
Host Species Mouse
Immunogen Pseudomonas aeruginosa serotype 6C
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.