Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudomonas Aeruginosa Antibody (95/159), Biotin, Novus Biologicals™
SDP

Catalog No. NB10064562B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064562B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064562B Supplier Novus Biologicals Supplier No. NB10064562B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Pseudomonas Aeruginosa
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 95/159
Conjugate Biotin
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias Pseudomonas aeruginosa 9
Host Species Mouse
Immunogen Pseudomonas aeruginosa, serotype 9 (NCTC 11448, ATCC 33356).
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for Pseudomonas aeruginosa, serotype 9.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.