Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudomonas Aeruginosa Antibody (95/159), FITC, Novus Biologicals™
SDP

Catalog No. NB213007 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB213007 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB213007 Supplier Novus Biologicals Supplier No. NB10064562F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pseudomonas Aeruginosa Monoclonal antibody specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen Pseudomonas Aeruginosa
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 95/159
Conjugate FITC
Formulation PBS
Gene Alias Pseudomonas aeruginosa 9
Host Species Mouse
Immunogen Pseudomonas aeruginosa, serotype 9 (NCTC 11448, ATCC 33356).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.