Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudomonas Aeruginosa Serotype 2B Antibody (1253/17), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1062876A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1062876A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1062876A70 Supplier Novus Biologicals Supplier No. NB10062876AF750

Mouse Monoclonal Antibody

Pseudomonas Aeruginosa Serotype 2B Monoclonal specifically detects Pseudomonas Aeruginosa Serotype 2B in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Pseudomonas Aeruginosa Serotype 2B
Applications ELISA
Classification Monoclonal
Clone 1253/17
Conjugate Alexa Fluor 750
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Pseudomonas aeruginosa serotype 2B
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This product is specific for P. aeruginosa serotype 2B.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.