Learn More
PSF/SFPQ Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00304

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Panda,
SFPQ, also named PSF, encodes a nuclear factor implicated in the splicing and regulation of gene expression. SFPQ probably forms a heteromer with NONO and participates in DNA pairing and DNA break repair program. Very recently SFPQ was identified as a downstream target of tau, complete nuclear depletion and cytoplasmic accumulation of SFPQ were shown in the neurons and astrocytes of brains with Alzheimer′s disease (AD), more strikingly, reduced SFPQ levels may progress together with tau pathology, these observation strongly suggests the important role of SFPQ pathology in neurodegenerative diseases including AD. SFPQ protein may appears as 47 kDa, 52 kDa or 100 kDa molecules in various cell types.Specifications
PSF/SFPQ | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P23246, Q8VIJ6 | |
SFPQ | |
Between 657 and 707 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
SFPQ | |
POMP100, PSF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6421, 71514 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.