Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSF1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290150 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290150 Supplier Novus Biologicals Supplier No. NBP230365PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSF1 Polyclonal antibody specifically detects PSF1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen PSF1
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias GINS complex subunit 1, GINS complex subunit 1 (Psf1 homolog), KIAA0186PSF1DNA replication complex GINS protein PSF1, partner of sld five-1
Host Species Rabbit
Immunogen Maps to a region between residue 1 to 50 of human GINS Complex Subunit 1 using the numbering given in entry NP_066545.3
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9837
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.