Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSG6 Antibody (935973), DyLight 650, R&D Systems™
SDP

Catalog No. MAB85981C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB85981C 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB85981C Supplier Novus Biologicals Supplier No. MAB85981C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSG6 Monoclonal specifically detects PSG6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen PSG6
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 935973
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CGM3, pregnancy specific beta-1-glycoprotein 6, Pregnancy-specific beta-1-glycoprotein 10, Pregnancy-specific beta-1-glycoprotein 12, pregnancy-specific beta-1-glycoprotein 6, Pregnancy-specific glycoprotein 10, Pregnancy-specific glycoprotein 12, Pregnancy-specific glycoprotein 6, PS-beta-G-10, PS-beta-G-12, PS-beta-G-6, PSBG-10, PSBG-12, PSBG-6, PSG10, PSG12, PSGGB
Gene Symbols PSG6
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived transfected with human PSG-6
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5675
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.