Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSGL-1/CD162 Antibody (PSGL1/1601), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB008117 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008117 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008117 Supplier Novus Biologicals Supplier No. NBP254380DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PSGL-1/CD162
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PSGL1/1601
Conjugate DyLight 594
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA
Gene Symbols SELPLG
Host Species Mouse
Immunogen Recombinant human PSGL-1/CD162 protein (Uniprot: Q14242)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.