Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMA/FOLH1/NAALADase I Antibody (GCP-04), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP1407A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP1407A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP1407A30 Supplier Novus Biologicals Supplier No. NBP145057AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMA/FOLH1/NAALADase I Monoclonal antibody specifically detects PSMA/FOLH1/NAALADase I in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen PSMA/FOLH1/NAALADase I
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GCP-04
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase
Gene Symbols FOLH1
Host Species Mouse
Immunogen Recombinant fragment of human GCPII (amino acids 44-750) produced in S2 cells (NM_001014986.1).
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2346
Test Specificity The antibody GCP-04 recognizes amino acids 100-104 of extracellular domain of denaturated glutamate carboxypeptidase II (PSMA, NAALADase, FOLH1), an approximately 95-110 kDa transmembrane glycoprotein.
Target Species Human, Mouse, Rat, Pig, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.