Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMA/FOLH1/NAALADase I Antibody (GCP-05), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N145058P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N145058P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N145058P594 Supplier Novus Biologicals Supplier No. NBP145058PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMA/FOLH1/NAALADase I Monoclonal specifically detects PSMA/FOLH1/NAALADase I in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PSMA/FOLH1/NAALADase I
Applications Flow Cytometry
Classification Monoclonal
Clone GCP-05
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase
Gene Symbols FOLH1
Host Species Mouse
Immunogen amino acids 44-750 of human GCPII
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2346.0
Test Specificity The antibody GCP-05 recognizes extracellular domain of glutamate carboxypeptidase II (NAALADase, FOLH1, PSMA), an approximately 95-110 kDa transmembrane glycoprotein expressed mainly in tumour neovasculatures, nervous system and jejunum, which is an important prostate tumour marker.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.