Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | PSMA4 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PSMA4 Polyclonal specifically detects PSMA4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PSMA4 | |
Polyclonal | |
Rabbit | |
Proteases & Other Enzymes | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
HC9EC 3.4.25.1, HsT17706, Macropain subunit C9, MGC12467, MGC24813, Multicatalytic endopeptidase complex subunit C9, proteasome (prosome, macropain) subunit, alpha type, 4, Proteasome component C9, proteasome subunit alpha type-4, proteasome subunit HC9, Proteasome subunit L, PSC9MGC111191 | |
PSMA4 | |
IgG | |
Affinity Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
P25789 | |
5685 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MEAIGHAGTCLGILANDGVLLAAERRNIHKLLDEVFFSEKIYKLNEDMACSVAGITSDANVLTNELRLIAQRYLLQYQEPIPCEQLVTALCDIKQA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title