Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMA7 Antibody (OTI3F11), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N273693A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273693A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273693A405 Supplier Novus Biologicals Supplier No. NBP273693AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMA7 Monoclonal specifically detects PSMA7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen PSMA7
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI3F11
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C6, EC 3.4.25.1, HSPCMGC3755, proteasome (prosome, macropain) subunit, alpha type, 7, proteasome subunit alpha 4, proteasome subunit alpha type-7, Proteasome subunit RC6-1, Proteasome subunit XAPC7, RC6-1, XAPC7
Gene Symbols PSMA7
Host Species Mouse
Immunogen Full length recombinant protein of human PSMA7 (NP_002783) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Core ESC Like Genes, Hypoxia, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5688.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.