Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219983 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219983 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219983 Supplier Novus Biologicals Supplier No. NBP219983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSMB5 Polyclonal specifically detects PSMB5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PSMB5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.67mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 1XPBS, 1% BSA, 10% Glycerol (pH7). with 0.01% Thimerosal
Gene Accession No. P28074
Gene Alias LMPXproteasome beta 5 subunit, Macropain epsilon chain, MB1EC 3.4.25.1, Multicatalytic endopeptidase complex epsilon chain, proteasome (prosome, macropain) subunit, beta type, 5, proteasome catalytic subunit 3, Proteasome chain 6, Proteasome epsilon chain, proteasome subunit beta type-5, Proteasome subunit MB1, Proteasome subunit X, proteasome subunit, beta type, 5, PSX large multifunctional protease X, XMGC104214
Gene Symbols PSMB5
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PSMB5. The exact sequence is proprietary.
Molecular Weight of Antigen 28 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5693
Test Specificity Expected to cross react based on sequence identity: Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.