Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB7 Antibody (OTI1E1), Novus Biologicals™
SDP

Catalog No. NBP201832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP201832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP201832 Supplier Novus Biologicals Supplier No. NBP201832
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMB7 Monoclonal specifically detects PSMB7 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Proteasome 20S beta 7
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1E1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150
Gene Alias EC 3.4.25.1, Macropain chain Z, Multicatalytic endopeptidase complex chain Z, proteasome (prosome, macropain) subunit, beta type, 7, proteasome subunit alpha, proteasome subunit beta 7, proteasome subunit beta type-7, Proteasome subunit Z, PSMB7, Zproteasome catalytic subunit 2
Gene Symbols PSMB7
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 58-277 of human PSMB7(NP_002790) produced in E.coli.
Molecular Weight of Antigen 29.8 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5695
Target Species Human, Mouse, Rat, Monkey, Primate
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.