Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMC1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290455 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290455 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290455 Supplier Novus Biologicals Supplier No. NBP214857PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSMC1 Polyclonal antibody specifically detects PSMC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen PSMC1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 26S protease regulatory subunit 4, 26S proteasome AAA-ATPase subunit RPT2, MGC24583, P26S4, p56, proteasome (prosome, macropain) 26S subunit, ATPase, 1, proteasome 26S ATPase subunit 1, Proteasome 26S subunit ATPase 1, proteasome 26S subunit, ATPase, 1, S4MGC8541
Host Species Rabbit
Immunogen This antibody maps to a region between residue 390 and 440 of human Proteasome (prosome, macropain) Subunit, ATPase, 1 using the numbering given in entry NP_002793.2 (GeneID 5700).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5700
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.