Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMC3IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PSMC3IP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
PSMC3IP Polyclonal specifically detects PSMC3IP in Human samples. It is validated for Western Blot.Specifications
PSMC3IP | |
Polyclonal | |
Purified | |
RUO | |
DBD-interacting, GT198, GT198 alternative, homologous-pairing protein 2 homolog, HOP2, HUMGT198A, Nuclear receptor coactivator GT198, Proteasome 26S ATPase subunit 3-interacting protein, PSMC3 interacting protein, Tat-binding protein 1-interacting protein, TBP-1 interacting protein, TBP-1-interacting protein, TBPIPPSMC3-interacting protein | |
PSMC3IP | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
NP_057640 | |
29893 | |
Synthetic peptide directed towards the middle region of human PSMC3IPThe immunogen for this antibody is PSMC3IP. Peptide sequence RLKNIKAATNHVTPEEKEQVYRERQKYCKEWRKRKRMATELSDAILEGYP. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title