Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD11 Antibody (1F4), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB310133 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310133 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310133 Supplier Novus Biologicals Supplier No. NBP130252MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMD11 Monoclonal antibody specifically detects PSMD11 in Human, Mouse, C. elegans samples. It is validated for Western Blot, ELISA

Specifications

Antigen PSMD11
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1F4
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias MGC3844,26S proteasome regulatory subunit RPN6, p44.5,26S proteasome regulatory subunit 9, proteasome (prosome, macropain) 26S subunit, non-ATPase, 11,26S proteasome regulatory subunit p44.5, Rpn6,26S proteasome regulatory subunit S9, S9,26S proteasome non-ATPase regulatory subunit 11
Host Species Mouse
Immunogen Recombinant human PSMD11 (1-422aa) purified from E. coli (NP_002806).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5717
Target Species Human, Mouse, C. elegans
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.