Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD11 Antibody (2C7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB162310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162310 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB162310 Supplier Novus Biologicals Supplier No. NBP259484AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMD11 Monoclonal antibody specifically detects PSMD11 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen PSMD11
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 2C7
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias MGC3844,26S proteasome regulatory subunit RPN6, p44.5,26S proteasome regulatory subunit 9, proteasome (prosome, macropain) 26S subunit, non-ATPase, 11,26S proteasome regulatory subunit p44.5, Rpn6,26S proteasome regulatory subunit S9, S9,26S proteasome non-ATPase regulatory subunit 11
Host Species Mouse
Immunogen Recombinant human PSMD11 (1-422aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5717
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.