Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD11 Mouse anti-Human, Mouse, Clone: 2C7, Novus Biologicals™
SDP

Catalog No. NBP25948450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25948450 50 μL
NBP259484X 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25948450 Supplier Novus Biologicals Supplier No. NBP25948450UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMD11 Monoclonal specifically detects PSMD11 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PSMD11
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2C7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1081, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Formulation Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Alias MGC3844,26S proteasome regulatory subunit RPN6, p44.5,26S proteasome regulatory subunit 9, proteasome (prosome, macropain) 26S subunit, non-ATPase, 11,26S proteasome regulatory subunit p44.5, Rpn6,26S proteasome regulatory subunit S9, S9,26S proteasome non-ATPase regulatory subunit 11
Gene Symbols PSMD11
Host Species Mouse
Immunogen Recombinant human PSMD11 (1-422aa) purified from E. coli
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5717
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.