Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD5 Polyclonal Antibody, Invitrogen™

Catalog No. PA1975
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1975 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1975 Supplier Thermo Scientific Supplier No. PA1975

Rabbit Polyclonal Antibody

PA1-975 detects purified human proteasome 19S subunit S5b. PA1-975 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽55 kDa protein representing purified human proteasome 19S subunit S5b. PA1-975 immunizing peptide corresponds to amino acid residues 485-504 from human proteasome 19S subunit S5b protein.

Proteolytic degradation is critical to the maintece of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signalling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (∽31), barrel-shaped molecular machine with an apparent molecular weight of ∽2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman’s syndrome, and Liddle syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMD5
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene PSMD5
Gene Accession No. Q16401
Gene Alias RP11-27I1.5, KIAA0072, MGC23145, S5B
Gene Symbols PSMD5
Host Species Rabbit
Immunogen Synthetic Peptide: L(485) S E G P Y Y V K P V S T T A V E G A E(504)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5711
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.