Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219989 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219989 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219989 Supplier Novus Biologicals Supplier No. NBP219989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSMD8 Polyclonal specifically detects PSMD8 in Human samples. It is validated for Western Blot.

Specifications

Antigen PSMD8
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P48556
Gene Alias 26S proteasome non-ATPase regulatory subunit 8, 26S proteasome regulatory subunit p31, HIP6,26S proteasome regulatory subunit S14, HYPF, MGC1660, Nin1p, p3126S proteasome regulatory subunit RPN12, proteasome (prosome, macropain) 26S subunit, non-ATPase, 8, Rpn12, S14
Gene Symbols PSMD8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PSMD8. The exact sequence is proprietary.
Molecular Weight of Antigen 40 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5714
Test Specificity Human. Species reactivity based on sequence identity: Chimpanzee (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.