Learn More
Description
Specifications
Specifications
| Antigen | PSME4 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | proteasome (prosome, macropain) activator subunit 4 |
| Gene Symbols | PSME4 |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Proteasome (Prosome, Macropain) Activator subunit 4 using the numbering given in entry NP_055429.2 (GeneID 23198). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
