Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMF1 Antibody (OTI2D7), HRP, Novus Biologicals™
SDP

Catalog No. NBP273707H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273707H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273707H Supplier Novus Biologicals Supplier No. NBP273707H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMF1 Monoclonal specifically detects PSMF1 in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen PSMF1
Applications Western Blot
Classification Monoclonal
Clone OTI2D7
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias hPI31, PI31, proteasome (prosome, macropain) inhibitor subunit 1 (PI31), proteasome inhibitor hP131 subunit, proteasome inhibitor PI31 subunit
Gene Symbols PSMF1
Host Species Mouse
Immunogen Full length recombinant protein of human PSMF1 (NP_848693) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 9491.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.