Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSP94/MSMB Antibody, Novus Biologicals™
SDP

Catalog No. NB397384 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397384 25 μL
NBP233610 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397384 Supplier Novus Biologicals Supplier No. NBP23361025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Prostate Secretory Protein/PSP Polyclonal specifically detects Prostate Secretory Protein/PSP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Prostate Secretory Protein/PSP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P08118
Gene Alias beta-microseminoprotein, IGBFProstate secreted seminal plasma protein, immunoglobulin binding factor, Immunoglobulin-binding factor, microseminoprotein, beta-, MSP, MSPB, PN44Prostate secretory protein of 94 amino acids, prostatic secretory protein 94, PRPS, PRSP, PSP, PSP57, PSP94HPC13, PSP-94Seminal plasma beta-inhibin
Gene Symbols MSMB
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: LKGNKHPINSEWQTDNCETCTCYETEISCCTLVSTPVGYDKDNCQRIFKKEDCKYIVVEKKDPK
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4477
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.