Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSP94/MSMB Antibody (OTI3D5), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP273625G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273625G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273625G Supplier Novus Biologicals Supplier No. NBP273625G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSP94/MSMB Monoclonal specifically detects PSP94/MSMB in Human samples. It is validated for Western Blot.

Specifications

Antigen PSP94/MSMB
Applications Western Blot
Classification Monoclonal
Clone OTI3D5
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IGBF, MSMB, MSPB, PIP, PN44, PRPS, PSP57, PSP94
Gene Symbols MSMB
Host Species Mouse
Immunogen Purified prostate specific antigen (MSMB) from human seminal fluid.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4477.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.